Learn More
Medchemexpress LLC Ntrc-844 | 1811503-67-9 | 95.6% | 489.54 g/mol | C28H28FN3O4 | 25MG
SDP
Catalog No. 5000237223
Encompass_Preferred
NTRC-844 is a selective antagonist of neurotensin receptor type 2 (NTS2) used in analgesic animal research; it has a reported EC50 of 238 nM, molecular formula C28H28FN3O4, and molecular weight 489.54 g/mol. The compound is supplied as a solid with listed purity 95.6%; manufacturer-provided datasheet, certificate of analysis, and safety data sheet are available. Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Selective NTS2 antagonist with reported EC50 of 238 nM.
  • Research-grade solid with listed purity of 95.6%.
  • Molecular formula C28H28FN3O4 and molecular weight 489.54 g/mol.
  • Multiple milligram-scale pack sizes available for research use.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Defined storage recommendations for powder and solutions.
Catalog No. 50-002-37223 Supplier Medchemexpress LLC Supplier No. HY12483125MG
NTRC-844 is a selective antagonist of neurotensin receptor type 2 (NTS2) used in analgesic animal research; it has a reported EC50 of 238 nM, molecular formula C28H28FN3O4, and molecular weight 489.54 g/mol. The compound is supplied as a solid with listed purity 95.6%; manufacturer-provided datasheet, certificate of analysis, and safety data sheet are available. Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Selective NTS2 antagonist with reported EC50 of 238 nM.
  • Research-grade solid with listed purity of 95.6%.
  • Molecular formula C28H28FN3O4 and molecular weight 489.54 g/mol.
  • Multiple milligram-scale pack sizes available for research use.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Defined storage recommendations for powder and solutions.