Learn More
Sigma Aldrich Fine Chemicals Biosciences Ntrc acd ACS reagent 70500ML
Catalog No. 502898270
Encompass
Nitric acid (HNO3) is a highly corrosive mineral acid. It is usually utilized for abstracting transition metal catalyst from single-walled carbon nanotubes (SWNTs) in order to purify them. The effect of nitric acid on the oxidation of multiwalled carbon nanotubes (MWCNTs) has been investigated by sample weight Raman spectrum solubility morphology and alignment examinations.
Catalog No. 50-289-8270 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 438073500ML
Nitric acid (HNO3) is a highly corrosive mineral acid. It is usually utilized for abstracting transition metal catalyst from single-walled carbon nanotubes (SWNTs) in order to purify them. The effect of nitric acid on the oxidation of multiwalled carbon nanotubes (MWCNTs) has been investigated by sample weight Raman spectrum solubility morphology and alignment examinations.