Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human NTRK1 (TRKA), His Tag Recombinant Protein

Catalog No. PR4672D
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR4672D 100 μg
PV3144 10 μg
PV4114 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR4672D Supplier Invitrogen™ Supplier No. PR4672D
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Cytoplasmic.

NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04629
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.2mM EDTA, 100mM NaCl, 5mM DTT, 50% glycerol, 0.05% Triton X-100 and no preservative; pH 7.5
Gene ID (Entrez) 4914
Molecular Weight (g/mol) 42.8 kDa
Name Human NTRK1 (TRKA), His Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias C80751; DKFZp781I14186; EC 2.7.10.1; gp140trk; high affinity nerve growth factor receptor; kinase TrkA; MTC; neurotrophic receptor tyrosine kinase 1; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Ntrk1; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; Tkr; TRK; TRK1; TRK1-transforming tyrosine kinase protein; Trka; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A
Common Name TrkA
Gene Symbol Ntrk1
Kinase Family Neurotrophic Receptor (NTRK) Kinase Family
Protein Length 441-796
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence 441-796
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Tyrosine Kinases
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Cytoplasmic
Form Liquid
Research Category Signal Transduction
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.