Learn More
Aviva Systems Biology Corp NTRK2 Antibody - C-terminal region (ARP51316_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51316_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NTRK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NTRK2. Alternate names: GP145-TrkB, TRKB, trk-BApplicationIF, WB, IHC. Species reactivity: Cow, Horse, Human, Rabbit, Rat
Catalog No. 50-285-575