Learn More
MSC Honeywell Safety Flextril 231 Chemical Resistant Gloves, Nitrile-Coated, ANSI Cut A3, Size XL, 12 Pairs
Catalog No. 5000508134
Encompass
FLEXTRIL™ 231 chemical resistant gloves offer advanced protection, combining a robust nitrile barrier with an HPPE-Nylon liner to provide cut and chemical resistance in demanding environments. These supported gloves are engineered with a rough texture to ensure a secure grip and feature Moisture Dispersion Technology to keep hands dry, making them suitable for applications in the automotive industry.
  • Provides ANSI/ISEA A3 cut resistance and EN 388 4X32C protection
  • Features a 1.7 mm thick nitrile coating for chemical resistance
  • Includes moisture dispersion technology for enhanced wearer comfort
  • Designed with rough texture for superior grip in wet and dry conditions
  • Tested for food contact compliance under FDA 21CFR.177.2600 and Commission Regulations (EU) No 10/2011
  • Constructed as a supported, powder-free, and silicone-free glove
Catalog No. 50-005-08134 Supplier MSC Supplier No. 18213983
FLEXTRIL™ 231 chemical resistant gloves offer advanced protection, combining a robust nitrile barrier with an HPPE-Nylon liner to provide cut and chemical resistance in demanding environments. These supported gloves are engineered with a rough texture to ensure a secure grip and feature Moisture Dispersion Technology to keep hands dry, making them suitable for applications in the automotive industry.
  • Provides ANSI/ISEA A3 cut resistance and EN 388 4X32C protection
  • Features a 1.7 mm thick nitrile coating for chemical resistance
  • Includes moisture dispersion technology for enhanced wearer comfort
  • Designed with rough texture for superior grip in wet and dry conditions
  • Tested for food contact compliance under FDA 21CFR.177.2600 and Commission Regulations (EU) No 10/2011
  • Constructed as a supported, powder-free, and silicone-free glove