Learn More
MSC Ansell MICROFLEX Supreno SE Nitrile Gloves, Powder-Free, X-Large, 100/Bx
Catalog No. 501909536
Encompass
MICROFLEX Supreno SE SU-690 gloves are engineered with a robust nitrile formulation to provide durable personal protection. These powder-free gloves feature textured fingertips for an assured grip in various conditions and offer advanced barrier protection with a low 0.65 AQL.
  • Robust nitrile formulation for durable personal protection
  • Advanced barrier protection with low 0.65 AQL
  • Textured fingertips for assured grip
  • Not made from natural rubber latex
  • Antistatic properties
  • Standard cuff length with beaded cuff
This item is not returnable. View return policy
Catalog No. 50-190-9536 Supplier MSC Supplier No. 70892328
MICROFLEX Supreno SE SU-690 gloves are engineered with a robust nitrile formulation to provide durable personal protection. These powder-free gloves feature textured fingertips for an assured grip in various conditions and offer advanced barrier protection with a low 0.65 AQL.
  • Robust nitrile formulation for durable personal protection
  • Advanced barrier protection with low 0.65 AQL
  • Textured fingertips for assured grip
  • Not made from natural rubber latex
  • Antistatic properties
  • Standard cuff length with beaded cuff