Learn More
Enzo Life Sciences Neutrophil elastase colorimetric drug discovery kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences BMLAK4970001
Encompass_Preferred
It is a complete assay system designed to screen inhibitors of neutrophil elastase, providing enough material to perform at least 96 assays. Neutrophil elastase, also known as leukocyte elastase (LE), is a serine protease found in polymorphonuclear neutrophils (PMNs) that functions in inflammation. It can cleave a variety of proteins, including fibrous elastin, collagen, clotting factors, TNF-α, and α1-antichymotrypsin. It has been shown to be taken up by tumor cells and accelerate tumor growth through its degradation of insulin receptor substrate-1 (IRS-1). Because of its potential roles in cancer and inflammatory diseases such as emphysema, cystic fibrosis, and atherosclerosis, it is a target for drug discovery. The kit uses the chromogenic substrate MeOSuc-Ala-Ala-Pro-Val-pNA; cleavage of p-nitroaniline (pNA) from the substrate increases absorbance at 405nm. Applications: Colorimetric detection, HTS Activity assay.
Catalog No. 50-201-0282