Learn More
Aviva Systems Biology Corp NTSR2 Antibody - C-terminal region (ARP64147_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64147_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NTSR2. It was validated on Western Blot. . Alternate names: NTR2ApplicationWB. Species reactivity: Horse, Human, Yeast
Catalog No. 50-288-187