Learn More
Bellco Glass, Inc. CO2 Tank Switch,230V
SDP
Catalog No. 501945837
Encompass_Preferred
Provides automatic switching from the main tank to auxiliary tank.. Rear panel hose barbs are for gas connection. Alarm will sound when tank pressure drops below 5psi. Manual switching is done via the button labeled tank switch. Mounts on top of any stand. MADE IN THE USA
Catalog No. 50-194-5837 Supplier Bellco Glass, Inc. Supplier No. 672700230
Provides automatic switching from the main tank to auxiliary tank.. Rear panel hose barbs are for gas connection. Alarm will sound when tank pressure drops below 5psi. Manual switching is done via the button labeled tank switch. Mounts on top of any stand. MADE IN THE USA