Learn More
Nuaire Inc NU-5820 INCUBATOR 200L CO2
SDP
Catalog No. NC1947148
Encompass
In-VitroCell ES NU-5820 Direct Heat 7 ft3 (200L) CO2 Incubator with Humidity ControlThe In-VitroCell ES NU-5820 Microbiological CO2 Incubator features relative humidity (RH) control for cell lines prone to desiccation by utilizing a sensor that measures humidity in the chamber and injects 99.99% HEPA filtered vapor into the growth chamber to maintain setpoint.
Catalog No. NC1947148 Supplier Nuaire Inc Supplier No. NU5820
Add to Cart
Add to Cart
Request a Quote
In-VitroCell ES NU-5820 Direct Heat 7 ft3 (200L) CO2 Incubator with Humidity ControlThe In-VitroCell ES NU-5820 Microbiological CO2 Incubator features relative humidity (RH) control for cell lines prone to desiccation by utilizing a sensor that measures humidity in the chamber and injects 99.99% HEPA filtered vapor into the growth chamber to maintain setpoint.