Learn More
MSC Nu-Calgon Acid Based Coil Cleaner, Liquid, 1 gal
Catalog No. 5000504512
Encompass
Cal-Brite® is a low-pH or acid-based product developed for cleaning evaporator coils, air-cooled condensers, and other finned cooling and heating coils. It is based upon glycolic acid and ammonium bifluoride. The activity between the glycolic acid and the ammonium bifluoride produces a synergistic cleaning effect, resulting in a clean, brightened coil but with the utmost in safety to the applicator and the job site as compared to HF-based products.
  • Foams and brightens
  • Safe for use on indoor and outdoor coils
  • Synergistic acid-based formulation
  • High hard water tolerance
  • Removes oxidation and brightens
  • Safer and less corrosive than HF-based cleaners
Catalog No. 50-005-04512 Supplier MSC Supplier No. 05423660
Cal-Brite® is a low-pH or acid-based product developed for cleaning evaporator coils, air-cooled condensers, and other finned cooling and heating coils. It is based upon glycolic acid and ammonium bifluoride. The activity between the glycolic acid and the ammonium bifluoride produces a synergistic cleaning effect, resulting in a clean, brightened coil but with the utmost in safety to the applicator and the job site as compared to HF-based products.
  • Foams and brightens
  • Safe for use on indoor and outdoor coils
  • Synergistic acid-based formulation
  • High hard water tolerance
  • Removes oxidation and brightens
  • Safer and less corrosive than HF-based cleaners