Learn More
MSC Nu-Calgon Bacteriostat Fungistat, Chlorine Dioxide, HVACR, EPA Registered, 1 gal.
Catalog No. 5000505188
Encompass
Bio-Fresh cd is a ready-to-use bacteriostat, fungistat, and deodorizer utilizing chlorine dioxide technology. It is specifically formulated and EPA registered for the control of bacteria, mold, mildew, fungi, and yeast in HVACR systems and Indoor Air Quality (IAQ) applications.
  • Concentrated, ready-to-use bacteriostat, fungistat, and deodorant
  • EPA registered for HVACR systems (No. 9804-3-65516)
  • Can be sprayed in occupied areas due to low toxicity
  • Eliminates odors by oxidizing the source, not masking
  • Inhibits microbial growth by reacting with cell walls
  • Water-based, non-flammable formulation
Catalog No. 50-005-05188 Supplier MSC Supplier No. 05423793
Bio-Fresh cd is a ready-to-use bacteriostat, fungistat, and deodorizer utilizing chlorine dioxide technology. It is specifically formulated and EPA registered for the control of bacteria, mold, mildew, fungi, and yeast in HVACR systems and Indoor Air Quality (IAQ) applications.
  • Concentrated, ready-to-use bacteriostat, fungistat, and deodorant
  • EPA registered for HVACR systems (No. 9804-3-65516)
  • Can be sprayed in occupied areas due to low toxicity
  • Eliminates odors by oxidizing the source, not masking
  • Inhibits microbial growth by reacting with cell walls
  • Water-based, non-flammable formulation