Learn More
MSC Nu-Calgon Coil Cleaner, Alkaline, 1 gal, Green
Catalog No. 5000508472
Encompass
This eco-friendly, low-foaming, emulsifying coil cleaner is formulated with natural ingredients. It quickly removes dust, dirt, and debris from evaporator coils, suitable for residential and light commercial applications. The product meets EPA Safer Choice Direct Release Criteria and is biodegradable, ensuring it will not harm plants, animals, or aquatic life.
  • Meets EPA Safer Choice Direct Release Criteria
  • Conforms to OEM specifications for cleaning microchannel coils
  • Ideal for any evaporator design
  • Low foaming, emulsifying coil cleaner
  • Perfect for residential and light commercial applications
  • Made with natural ingredients and eco-friendly resources
  • Biodegradable and will not harm plants, animals, or aquatic life
  • Effectively removes dust, dirt, and debris from evaporator coils
Catalog No. 50-005-08472 Supplier MSC Supplier No. 62410121
This eco-friendly, low-foaming, emulsifying coil cleaner is formulated with natural ingredients. It quickly removes dust, dirt, and debris from evaporator coils, suitable for residential and light commercial applications. The product meets EPA Safer Choice Direct Release Criteria and is biodegradable, ensuring it will not harm plants, animals, or aquatic life.
  • Meets EPA Safer Choice Direct Release Criteria
  • Conforms to OEM specifications for cleaning microchannel coils
  • Ideal for any evaporator design
  • Low foaming, emulsifying coil cleaner
  • Perfect for residential and light commercial applications
  • Made with natural ingredients and eco-friendly resources
  • Biodegradable and will not harm plants, animals, or aquatic life
  • Effectively removes dust, dirt, and debris from evaporator coils