Learn More
MSC Nu-Calgon Coil Cleaner, Alkaline, No-Rinse, Biodegradable, 1 gal
Catalog No. 5000505145
Encompass
Evap Pow'r-C is a no-rinse, green-colored, slightly alkaline liquid coil cleaner designed for cleaning indoor evaporator coils. This metal-safe and biodegradable product cleans and emulsifies stubborn soils and deposits. It is NSF A1/K4 Registered for safe use in food processing.
  • No-rinse coil cleaning
  • Cleans and emulsifies stubborn soils and deposits
  • Metal safe; will not harm evaporator coil's metal surfaces
  • Alkaline, near neutral pH formulation
  • Biodegradable
  • NSF A1/K4 registered for safe food processing areas
  • Restores lost capacity and efficiencies of coils
  • Avoids corrosion risks associated with aggressive or corrosive cleaners
Catalog No. 50-005-05145 Supplier MSC Supplier No. 05423728
Evap Pow'r-C is a no-rinse, green-colored, slightly alkaline liquid coil cleaner designed for cleaning indoor evaporator coils. This metal-safe and biodegradable product cleans and emulsifies stubborn soils and deposits. It is NSF A1/K4 Registered for safe use in food processing.
  • No-rinse coil cleaning
  • Cleans and emulsifies stubborn soils and deposits
  • Metal safe; will not harm evaporator coil's metal surfaces
  • Alkaline, near neutral pH formulation
  • Biodegradable
  • NSF A1/K4 registered for safe food processing areas
  • Restores lost capacity and efficiencies of coils
  • Avoids corrosion risks associated with aggressive or corrosive cleaners