Learn More
MSC Nu-Calgon Coil Gun, 2-Quart, 5 Mix Ratios, Quick-Disconnect, 1 each
Catalog No. 5000505568
Encompass
The Coil Gun is designed for spraying various coil cleaners for cleaning air-cooled condensers, evaporator coils, and similar surfaces. It automatically blends chemical and water in the mixing head during spraying, eliminating the need for pre-mixing.
  • Quick-disconnect feature
  • Five mix ratio settings: 3:1, 4:1, 6:1, 9:1 & 10:1
  • Has a 2-quart capacity
  • Foam wand to enhance foaming
  • Deflector tip to fan out spray
  • Connects to optional probe accessory for 90-degree spray
  • Recommended water pressure: 40 to 60 psi
  • Built-in anti-siphon device
  • Solid stream spray distance up to 25 ft.
Catalog No. 50-005-05568 Supplier MSC Supplier No. 07331051
The Coil Gun is designed for spraying various coil cleaners for cleaning air-cooled condensers, evaporator coils, and similar surfaces. It automatically blends chemical and water in the mixing head during spraying, eliminating the need for pre-mixing.
  • Quick-disconnect feature
  • Five mix ratio settings: 3:1, 4:1, 6:1, 9:1 & 10:1
  • Has a 2-quart capacity
  • Foam wand to enhance foaming
  • Deflector tip to fan out spray
  • Connects to optional probe accessory for 90-degree spray
  • Recommended water pressure: 40 to 60 psi
  • Built-in anti-siphon device
  • Solid stream spray distance up to 25 ft.