Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Nu-Calgon Ice Machine Cleaner, Phosphoric Acid, Liquid, 1 gal

Catalog No. 5000504289
Encompass

This liquid ice machine cleaner is formulated with phosphoric acid for effective descaling and maintenance of various ice machine types.

  • Container type: Bottle
  • Container size: 1 gal
  • Formula type: Phosphoric acid
  • Applications include: Cube, tube, flake, and commercial dishwasher ice machines

Catalog No. 50-005-04289 Supplier MSC Supplier No. 07328768
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This liquid ice machine cleaner is formulated with phosphoric acid for effective descaling and maintenance of various ice machine types.

  • Container type: Bottle
  • Container size: 1 gal
  • Formula type: Phosphoric acid
  • Applications include: Cube, tube, flake, and commercial dishwasher ice machines

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.