Rx11-flush is a unique HFC-based solvent engineered for flushing refrigeration and air conditioning systems. It efficiently flushes away sludge, carbon residue, oils, acids, water, and other particulates. The product is non-toxic, non-flammable, and non-ozone depleting, making it ideal for system clean-up after burnouts and R-410A conversions.
- Unique HFC-based solvent formulation
- Non-toxic, non-flammable, and non-ozone depleting
- A2L ready
- Industry standard for flushing refrigeration and air conditioning systems
- Effectively flushes line sets for R-410A conversions and retrofits
- Dries quickly with low boiling point for complete evaporation
- Will not attack system components
- Approved under U.S. EPA's SNAP Program
- Flushes systems up to 14 tons
- Includes 2x 2 lb cans and 1 injection valve