Learn More
MSC Nu-Calgon Vacuum Pump Oil, Multi-Viscosity, Low Vapor Pressure, 1 gal
Catalog No. 5000505262
Encompass
Nu-Calgon Vacuum Pump Oil is a high-grade lubricant blend designed for deep vacuum performance. It lubricates to minimize wear, provides surface protection, and maintains a high viscosity index for effective operation across varying pressures and temperatures. This oil is essential for the evacuation of moisture and contaminants from refrigeration and air conditioning systems, ensuring optimal vacuum pump performance.
  • Multi-viscosity oil with a viscosity index of 100
  • Provides peak performance across a wide range of temperatures and pressures
  • Lubricates and seals effectively at operating temperatures developed by the pump
  • Very low vapor pressure, a critical characteristic for vacuum pump oil
  • Suitable for evacuation of moisture and contaminants
Catalog No. 50-005-05262 Supplier MSC Supplier No. 07329360
Nu-Calgon Vacuum Pump Oil is a high-grade lubricant blend designed for deep vacuum performance. It lubricates to minimize wear, provides surface protection, and maintains a high viscosity index for effective operation across varying pressures and temperatures. This oil is essential for the evacuation of moisture and contaminants from refrigeration and air conditioning systems, ensuring optimal vacuum pump performance.
  • Multi-viscosity oil with a viscosity index of 100
  • Provides peak performance across a wide range of temperatures and pressures
  • Lubricates and seals effectively at operating temperatures developed by the pump
  • Very low vapor pressure, a critical characteristic for vacuum pump oil
  • Suitable for evacuation of moisture and contaminants