Learn More
Selleck Chemical LLC NU2058 10mg 161058-83-9 O(6)-Cyclohexylmethylguanine
SDP
Catalog No. 501948447
Encompass_Preferred
NU2058 (O(6)-Cyclohexylmethylguanine) is an inhibitor of CDK2 with IC50 value of 17 ?M in an isolated enzyme assay. It also potentiates melphalan (DMF 2.3), and monohydroxymelphalan (1.7), but not temozolomide or ionising radiation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8447 Supplier Selleck Chemical LLC Supplier No. S531610MG
Add to Cart
Add to Cart
NU2058 (O(6)-Cyclohexylmethylguanine) is an inhibitor of CDK2 with IC50 value of 17 ?M in an isolated enzyme assay. It also potentiates melphalan (DMF 2.3), and monohydroxymelphalan (1.7), but not temozolomide or ionising radiation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.