Learn More
Enzo Life Sciences NU2058 (25mg). CAS: 161058-83-9
SDP
Supplier:  Enzo Life Sciences ALX270394M025
Encompass_Preferred
Competitive inhibitor of CDK1/cyclin B (IC50=5µM) and CDK2/cyclin A (IC50=12µM). Pharmacologically distinct from the relative specific inhibitor of CDK1/2 roscovitine (BML-CC205). Alternative name: O(6)-Cyclohexylmethylguanine. Solubility: Soluble in water or DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-8533