Learn More
Medchemexpress LLC HY-19316 5mg Medchemexpress, NU2058 CAS:161058-83-9 Purity:>98%
SDP
Catalog No. 502082112
Encompass_Preferred
Medchemexpress, HY-19316 5mg NU2058 CAS:161058-83-9 NU2058 (O6-(Cyclohexylmethyl)guanine) is a potent, competitive and guanine-based CDK inhibitor with IC50s of 17 μM and 26 μM for CDK2 and CDK1. NU2058 has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2112 Supplier Medchemexpress LLC Supplier No. HY193165MG
Medchemexpress, HY-19316 5mg NU2058 CAS:161058-83-9 NU2058 (O6-(Cyclohexylmethyl)guanine) is a potent, competitive and guanine-based CDK inhibitor with IC50s of 17 μM and 26 μM for CDK2 and CDK1. NU2058 has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.