Learn More
TARGETMOL CHEMICALS INC NU6102 1MG
SDP
Catalog No. 502580534
Encompass_Preferred
Also available in and bulk. Please contact Fisher for quotes. NU6102 is a selective and potent ATP-competitive CDK2 inhibitor with antitumor activity against CDK1/cyclinB CDK2/cyclinA3 CDK1/CDK2 CDK4 DYRK1A PDK1 and ROCKII and it can be used to study rectal cancer. purity: 99%
Catalog No. 50-258-0534 Supplier TARGETMOL CHEMICALS INC Supplier No. T282181MG
Also available in and bulk. Please contact Fisher for quotes. NU6102 is a selective and potent ATP-competitive CDK2 inhibitor with antitumor activity against CDK1/cyclinB CDK2/cyclinA3 CDK1/CDK2 CDK4 DYRK1A PDK1 and ROCKII and it can be used to study rectal cancer. purity: 99%