Learn More
Enzo Life Sciences NU6102 (5mg). CAS: 444722-95-6
SDP
Supplier:  Enzo Life Sciences ALX270419M005
Encompass_Preferred
1,000-fold more potent inhibitor of CDK1/cyclin B (IC50=9.5nM) and CDK2/cyclin A3 (IC50=5.4nM) than NU2058 (ALX-270-394). Selective for CDK1 and CDK2 compared to CDK4/D1 (IC50=1.6µM), DYRK1A (IC50=0.9µM), PDK1 (IC50=0.8µM) and ROCKII (IC50=0.6µM). Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8545