Learn More
AdipoGen NU6102 (5 mg)
SDP
Supplier:  AdipoGen AGCR10020M005
Encompass_Preferred
Chemical. CAS: 444722-95-6. Formula: C18H22N6O3S. MW: 402.5. Potent CDK1/cyclin B (IC50 = 9.5 nM) and CDK2/cyclin A3 (IC50 = 5.4 nM) inhibitor. 1'000-fold more potent than NU2058. Selective for CDK1 and CDK2 compared to CDK4/D1 (IC50 = 1.6 µM), DYRK1A (IC50 = 0.9 µM), PDK1 (IC50 = 0.8 µM) and ROCK-II (IC50 = 0.6 µM). Inhibits cell growth.
Catalog No. 50-205-3769