Learn More
Abcam NU6102, CDK1/cyclin B and CDK2/cyclin A3 inhibitor, 5MG
Catalog No. 502705885
Encompass_Preferred
MW 402.5 Da, Purity >98%. Potent CDK1/cyclin B and CDK2/cyclin A3 inhibitor (IC50 values are 9.5 and 5.4 nM for CDK1/cyclin B and CDK2/cyclin A3 respectively). Antiproliferative agent. Inhibits CDK4/D1, DYRK1A, PDK1 ROCKII (IC50 values are 1.6, 0.9, 0.8 and 0.6 μM respectively). Induces G2/M cell cycle arrest. Shows antitumor effects *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5885 Supplier Abcam Supplier No. AB1443175MG
MW 402.5 Da, Purity >98%. Potent CDK1/cyclin B and CDK2/cyclin A3 inhibitor (IC50 values are 9.5 and 5.4 nM for CDK1/cyclin B and CDK2/cyclin A3 respectively). Antiproliferative agent. Inhibits CDK4/D1, DYRK1A, PDK1 ROCKII (IC50 values are 1.6, 0.9, 0.8 and 0.6 μM respectively). Induces G2/M cell cycle arrest. Shows antitumor effects *in vivo.*.
The product is subject to the following: Abcam Restricted Use Statement