Learn More
Medchemexpress LLC HY-107419 5mg Medchemexpress, NU6140 CAS:444723-13-1 Purity:>98%
SDP
Catalog No. 501932284
Encompass_Preferred
Medchemexpress, HY-107419 5mg NU6140 CAS:444723-13-1 NU6140 is a selective CDK2-cyclin A inhibitor ( IC 50 , 0.41 μM), exhibits 10- to 36-fold selectivity over other CDKs [1] . NU6140 also potently inhibits Aurora A and Aurora B , with IC 50 s of 67 and 35 nM, respectively [2] . Enhances the apoptotic effect, with anti-cancer activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2284 Supplier Medchemexpress LLC Supplier No. HY1074195MG
Medchemexpress, HY-107419 5mg NU6140 CAS:444723-13-1 NU6140 is a selective CDK2-cyclin A inhibitor ( IC 50 , 0.41 μM), exhibits 10- to 36-fold selectivity over other CDKs [1] . NU6140 also potently inhibits Aurora A and Aurora B , with IC 50 s of 67 and 35 nM, respectively [2] . Enhances the apoptotic effect, with anti-cancer activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.