Learn More
Selleck Chemical LLC NU7026 10mg 154447-35-5 LY293646
SDP
Supplier:  Selleck Chemical LLC S289310MG
Encompass_Preferred
NU7026 (LY293646) is a potent DNA-PK inhibitor with IC50 of 0.23 ?M in cell-free assays, 60-fold selective for DNA-PK than PI3K and inactive against both ATM and ATR. NU7026 enhances G2/M cell arrest and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0513443