Learn More
Cayman Chemical NU 7441, 503468-95-9, 5 mg
SDP
Supplier:  Cayman Chemical 148815
Encompass_Preferred
A selective DNA-PK inhibitor (IC50 = 14 nM) that increases the cytotoxicity of ionizing radiation and etoposide in human colon cancer cell lines in vitro and potentiates the effects of etoposide in mice bearing human colon cancer xenograft tumors in vivo
Catalog No. NC1523574