Learn More
Selleck Chemical LLC NU7441 (KU-57788) S2638-200mg
SDP
Catalog No. 503111760
Encompass_Preferred
NU7441 (KU-57788) is a highly potent and selective DNA-PK inhibitor with IC50 of 14 nM and also inhibits mTOR and PI3K with IC50 of 1 7 M and 5 M in cell-free assays respectively It reduces the frequency of NHEJ while increasing the rate of HDR following Cas9-mediated DNA cleavage
Catalog No. 50-311-1760 Supplier Selleck Chemical LLC Supplier No. S2638200MG
NU7441 (KU-57788) is a highly potent and selective DNA-PK inhibitor with IC50 of 14 nM and also inhibits mTOR and PI3K with IC50 of 1 7 M and 5 M in cell-free assays respectively It reduces the frequency of NHEJ while increasing the rate of HDR following Cas9-mediated DNA cleavage