Learn More
Apexbio Technology LLC NU7441 (KU-57788), 503468-95-9, MFCD11983069, 5 mg
SDP
Catalog No. 501013746
Encompass_Preferred
MF: C25H19NO3S, MW: 413.49, Solubility: 4.13mg/mL in DMSO, insoluble in EtOH,insoluble in H2O. NU7441 is a selective inhibitor of DNA-dependent protein kinase (DNA-PK) with IC50 value of 13 nM.
Catalog No. 50-101-3746 Supplier Apexbio Technology LLC Supplier No. A83155
Add to Cart
Add to Cart
MF: C25H19NO3S, MW: 413.49, Solubility: 4.13mg/mL in DMSO, insoluble in EtOH,insoluble in H2O. NU7441 is a selective inhibitor of DNA-dependent protein kinase (DNA-PK) with IC50 value of 13 nM.