Learn More
Medchemexpress LLC HY-110127 5mg , NU9056 CAS:1450644-28-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1101275MG
Encompass_Preferred
HY-110127 5mg NU9056 CAS: 1450644-28-6 NU9056 is a potent and selective Tip60 (KAT5) histone acetyltransferase inhibitor with an of 2 µM. NU9056 shows >16-fold selectivity for Tip60 over PCAF, p300 and GCN5. NU9056 induces apoptosis of prostate cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0518