Learn More
Sino Biological Recombinant Human NUAK2 Protein (Full Length, K81R, GST Tag)
Catalog No. 502568453
Encompass_Preferred
Recombinant full length human NUAK2 (K81R) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-256-8453 Supplier Sino Biological Supplier No. N2016G20
Recombinant full length human NUAK2 (K81R) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.