Learn More
Apexbio Technology LLC NUC/CYTO FRACTION KIT-100mg
SDP
Catalog No. 5000897384
Encompass_Preferred
Nuclear/Cytosol Fractionation Kit is a crucial tool in the field of biomedical research originating from advancements in cellular biology techniques It enables the separation of nuclear and cytosolic components facilitating the study of signaling pathways and protein localization Its mechanism involves selective lysis and centrifugation to isolate cellular fractions This kit is commonly used in in vitro studies often in the nanomolar to low micromolar range to investigate cellular processes drug responses and protein interactions Typical applications include cell model experiments animal studies and drug screening Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97384 Supplier Apexbio Technology LLC Supplier No. K2111100
Nuclear/Cytosol Fractionation Kit is a crucial tool in the field of biomedical research originating from advancements in cellular biology techniques It enables the separation of nuclear and cytosolic components facilitating the study of signaling pathways and protein localization Its mechanism involves selective lysis and centrifugation to isolate cellular fractions This kit is commonly used in in vitro studies often in the nanomolar to low micromolar range to investigate cellular processes drug responses and protein interactions Typical applications include cell model experiments animal studies and drug screening Experimental concentrations typically depend on the specific research design