Learn More
Aviva Systems Biology Corp NUCB1 Antibody - C-terminal region (P102230_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P102230_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NUCB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NUCB1. Alternate names: NUC, CALNUCApplicationWB. Species reactivity: Human
Catalog No. 50-014-53812