Learn More
Aviva Systems Biology Corp NUCB2 Antibody - middle region (ARP36567_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36567_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NUCB2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human NUCB2. Alternate names: NEFAApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-53053