Learn More
TARGETMOL CHEMICALS INC NUCC-390 5MG
SDP
Catalog No. 502581472
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NUCC-390 is a novel and selective small-molecule CXCR4 receptor agonist. NUCC-390 induces internalization of CXCR4 receptors and acts in an opposite way of AMD3100. NUCC-390 promotes nerve recovery of function after neurodegeneration in vivo. purity: 98%
Catalog No. 50-258-1472 Supplier TARGETMOL CHEMICALS INC Supplier No. T122695MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NUCC-390 is a novel and selective small-molecule CXCR4 receptor agonist. NUCC-390 induces internalization of CXCR4 receptors and acts in an opposite way of AMD3100. NUCC-390 promotes nerve recovery of function after neurodegeneration in vivo. purity: 98%