Learn More
Medchemexpress LLC Nucc-390 dihydrochloride | 2749281-71-6 | MFCD32690075 | 98.2% | 468.46 g·mol⁻¹ | C23H35Cl2N5O | 5 MG
SDP
Catalog No. 5000263521
Encompass_Preferred
NUCC-390 dihydrochloride is a small-molecule dihydrochloride salt that acts as a selective CXCR4 receptor agonist for research use. It induces CXCR4 internalization and has been reported to promote nerve recovery after neurodegeneration. The compound is supplied as a light yellow to yellow solid, with a molecular formula of C23H35Cl2N5O and a molecular weight of 468.46 g·mol⁻¹.
  • Selective CXCR4 receptor agonist that induces receptor internalization.
  • Reported to promote nerve recovery after neurodegeneration in vivo.
  • High purity suitable for research applications (98.2%).
  • Supplied as a solid; also available as a DMSO solution for convenience.
  • Stable when stored as recommended (powder at 4°C; in solvent -80°C or -20°C under nitrogen).
Catalog No. 50-002-63521 Supplier Medchemexpress LLC Supplier No. HY111793A5MG
NUCC-390 dihydrochloride is a small-molecule dihydrochloride salt that acts as a selective CXCR4 receptor agonist for research use. It induces CXCR4 internalization and has been reported to promote nerve recovery after neurodegeneration. The compound is supplied as a light yellow to yellow solid, with a molecular formula of C23H35Cl2N5O and a molecular weight of 468.46 g·mol⁻¹.
  • Selective CXCR4 receptor agonist that induces receptor internalization.
  • Reported to promote nerve recovery after neurodegeneration in vivo.
  • High purity suitable for research applications (98.2%).
  • Supplied as a solid; also available as a DMSO solution for convenience.
  • Stable when stored as recommended (powder at 4°C; in solvent -80°C or -20°C under nitrogen).