Learn More
Macherey-Nagel NucleoSpin Blood XL, 50 preps
Catalog No. NC1105387
Encompass_Preferred
NucleoSpin Blood XL (50) 50 preps for the isolation of genomic DNA from blood - NucleoSpin Blood XL Columns, Collection Tubes, buffers, Proteinase K
Catalog No. NC1105387 Supplier Macherey-Nagel Supplier No. 740950.50
NucleoSpin Blood XL (50) 50 preps for the isolation of genomic DNA from blood - NucleoSpin Blood XL Columns, Collection Tubes, buffers, Proteinase K