Learn More
New England Biolabs, Inc. Nuclease-free Water – 100 ml (4 x 25 ml)
SDP
Catalog No. 501526333
Encompass
Nuclease-free Water is ideal for the preparation of reagents and for use in enzymatic reactions. No toxic agents, such as DEPC, are used in the production of this water, so as to avoid inhibition in enzymatic reactions.
Catalog No. 50-152-6333 Supplier New England Biolabs, Inc. Supplier No. B1500L
Nuclease-free Water is ideal for the preparation of reagents and for use in enzymatic reactions. No toxic agents, such as DEPC, are used in the production of this water, so as to avoid inhibition in enzymatic reactions.