Learn More
Enzo Life Sciences Nuclear extract from HeLa cells (100µl)
SDP
Catalog No. 502011096
Encompass_Preferred
Prepared by high salt extraction of HeLa nuclei (human cervical cancer cell line), this extract is a rich source of HDAC activity, useful for inhibitor screening or immunoprecipitation of HDAC containing complexes. Typically, 0.5µl or less per well is sufficient for assay with the HDAC FLUOR DE LYS fluorometric activity assay (BML-AK500). Formulation: Liquid. In 0.1M KCl, 20mM HEPES/NaOH, pH 7.9, 20% (v/v) glycerol, 0.2mM EDTA, 0.5mM DTT, 0.5mM PMSF. Long Term Storage: -80°C.
Catalog No. 50-201-1096 Supplier Enzo Life Sciences Supplier No. BMLKI1400100
Add to Cart
Add to Cart
Prepared by high salt extraction of HeLa nuclei (human cervical cancer cell line), this extract is a rich source of HDAC activity, useful for inhibitor screening or immunoprecipitation of HDAC containing complexes. Typically, 0.5µl or less per well is sufficient for assay with the HDAC FLUOR DE LYS fluorometric activity assay (BML-AK500). Formulation: Liquid. In 0.1M KCl, 20mM HEPES/NaOH, pH 7.9, 20% (v/v) glycerol, 0.2mM EDTA, 0.5mM DTT, 0.5mM PMSF. Long Term Storage: -80°C.