Learn More
Abcam Nuclear E x traction Kit, 100 Tests
Catalog No. NC0555748
Encompass_Preferred
Nuclear extraction kit provides a simple and selective method for nuclear protein extraction and fractionation from cells or tissue samples in just 1 hour. This kit includes all necessary reagents to obtain high yields of nuclear proteins suitable for various downstream applications. - Cited in over 255 publications - High retention of enzyme activity – contains 80% less salt than conventional kits and no SDS, allowing better enzyme activity retention - Suitable for western blotting, protein-DNA binding assays, nuclear enzyme assays, and more - Fast, simple procedure that ensures optimized nuclear protein isolation
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0555748 Supplier Abcam Supplier No. AB113474100TEST
Nuclear extraction kit provides a simple and selective method for nuclear protein extraction and fractionation from cells or tissue samples in just 1 hour. This kit includes all necessary reagents to obtain high yields of nuclear proteins suitable for various downstream applications. - Cited in over 255 publications - High retention of enzyme activity – contains 80% less salt than conventional kits and no SDS, allowing better enzyme activity retention - Suitable for western blotting, protein-DNA binding assays, nuclear enzyme assays, and more - Fast, simple procedure that ensures optimized nuclear protein isolation
The product is subject to the following: Abcam Restricted Use Statement