Learn More
Abcam Nuclear E x traction Kit, 100 Tests
Supplier:  Abcam AB221978100TEST
Encompass_Preferred
This Nuclear Extraction Kit is formulated for the quick and simple isolation of nuclear and cytoplasmic fractions from cultured cells and tissue homogenates that can be used successfully in our transcription factor assay kits.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2170289