Learn More
Cayman Chemical Nuclear Extraction Kit, 1 ea
SDP
Catalog No. 502906910
Encompass_Preferred
Preparation of nuclear extracts is the first step in examining transcription factor activity. Cayman’s Nuclear Extraction is formulated for the quick and simple isolation of nuclear and cytoplasmic fractions from cultured cells and tissue homogenates that can be used successfully in Cayman’s Transcription Factor Assay kits. The proteins isolated using this kit can also be used in electrophoretic mobility shift assays (EMSA) and western blotting applications.
Catalog No. 50-290-6910 Supplier Cayman Chemical Supplier No. 100092771 EA
Preparation of nuclear extracts is the first step in examining transcription factor activity. Cayman’s Nuclear Extraction is formulated for the quick and simple isolation of nuclear and cytoplasmic fractions from cultured cells and tissue homogenates that can be used successfully in Cayman’s Transcription Factor Assay kits. The proteins isolated using this kit can also be used in electrophoretic mobility shift assays (EMSA) and western blotting applications.