Learn More
Cayman Chemical Nuclear Factr of ActIvated T
SDP
Supplier:  Cayman Chemical 3403896 WELL
Encompass_Preferred
Reporter cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields high cell viability post-thaw and provides the convenience of immediately dispensing healthy reporter cells into assay plates. There is no need for intermediate preparatory steps such as the spin-and-rinse of cells, viability determinations, cell titer adjustments, or the pre-incubation of reporter cells prior to assay setup. [INDIGO Catalog Nos. IB18001, IB18001-32, IB18002]
Catalog No. 50-294-7008