Learn More
Cayman Chemical Nuclear Factor of Activated T cells Reporter Assay System, 3x32 Well
SDP
Catalog No. 502947007
Encompass_Preferred
Reporter cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields high cell viability post-thaw and provides the convenience of immediately dispensing healthy reporter cells into assay plates. There is no need for intermediate preparatory steps such as the spin-and-rinse of cells, viability determinations, cell titer adjustments, or the pre-incubation of reporter cells prior to assay setup. [INDIGO Catalog Nos. IB18001, IB18001-32, IB18002]
Catalog No. 50-294-7007 Supplier Cayman Chemical Supplier No. 340383X32 WELL
Reporter cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryopreservation method yields high cell viability post-thaw and provides the convenience of immediately dispensing healthy reporter cells into assay plates. There is no need for intermediate preparatory steps such as the spin-and-rinse of cells, viability determinations, cell titer adjustments, or the pre-incubation of reporter cells prior to assay setup. [INDIGO Catalog Nos. IB18001, IB18001-32, IB18002]