Learn More
Vector Laboratories VECTOR Nuclear Fast Red 500 ml
SDP
Catalog No. NC9483816
Encompass
Nuclei stained with Vector Nuclear Fast Red have a pink appearance. This counterstain provides good contrast with a variety of substrates and is suitable for use with non-aqueous mountants. The staining procedure is rapid, requiring only one step. This counterstain provides an excellent alternative in IHC assays where hematoxylin obscures the substrate colors.
Catalog No. NC9483816 Supplier Vector Laboratories Supplier No. H3403500
Nuclei stained with Vector Nuclear Fast Red have a pink appearance. This counterstain provides good contrast with a variety of substrates and is suitable for use with non-aqueous mountants. The staining procedure is rapid, requiring only one step. This counterstain provides an excellent alternative in IHC assays where hematoxylin obscures the substrate colors.