Learn More
Diagnostic Biosystems NUCLEAR FAST RED FOR IHC Nuclear Fast Red
Catalog No. 502407444
Encompass_Preferred
Nuclear Fast Red is a histological staining reagent suitable for visualization of nuclei in tissue sections. Nuclear Fast Red provides a red counterstain designed for optimal performance when used in immunohistochemistry.\n\nDBS’s Nclear Fast Red is intended for use in the histologic demonstration of nuclear staining. This staining technique is used to make the critical distinction between a normal nucleus and an abnormal one. When used as a counterstain in immunohstochemical procedures, Nuclear Fast Red is compatible with most commonly used chromogens including DBS HRP-Yellow, HRP-Green, HRP-Black, and DAB. Nuclear Fast Red is particularly useful with blue chromogens, such as DBS HRP-Blue.
Catalog No. 50-240-7444 Supplier Diagnostic Biosystems Supplier No. K098
Nuclear Fast Red is a histological staining reagent suitable for visualization of nuclei in tissue sections. Nuclear Fast Red provides a red counterstain designed for optimal performance when used in immunohistochemistry.\n\nDBS’s Nclear Fast Red is intended for use in the histologic demonstration of nuclear staining. This staining technique is used to make the critical distinction between a normal nucleus and an abnormal one. When used as a counterstain in immunohstochemical procedures, Nuclear Fast Red is compatible with most commonly used chromogens including DBS HRP-Yellow, HRP-Green, HRP-Black, and DAB. Nuclear Fast Red is particularly useful with blue chromogens, such as DBS HRP-Blue.