Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclear Fast Red, Sigma-Aldrich

Catalog No. 01259020 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-020 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-020 Supplier MilliporeSigma Supelco Supplier No. 6070025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

suitable for microscopy

Nuclear Fast Red (C. l. 60760) is a substituted hydroxy anthraquinone chromophore that can chelate metal ions. It is also known as kernechtrot and 4-Amino-9,10-dihydro-1,3-dihydroxy-9,10-dioxo-2-anthracenesulfonic acid sodium salt. It is used for human-medical cell diagnosis and serves the purpose of the histological investigation of sample material of human origin. It is a dry staining dye that makes target structures (by fixing, wherenecessary embedding, staining with the above nuclear fast red solution, counterstaining, mounting) in human-histological specimen materials, for examplehistological sections of e. g. the kidney, evaluable for diagnostic purposes.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6409-77-4
Color Red
Quantity 25 g
Molecular Weight (g/mol) 357.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.