Learn More
Abcam Nuclear Fast Red Solution, 125ML
Catalog No. NC1976934
Encompass_Preferred
Nuclear fast red stain solution (pH 2 - 4) is a stain for histological applications. - Stains nuclei red and cytoplasm pale pink - Contains 80% water, 5% aluminium sulfate, 0.2% nuclear fast red - Enhanced stability - Prevents precipitation over time
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1976934 Supplier Abcam Supplier No. AB246831125ML
Nuclear fast red stain solution (pH 2 - 4) is a stain for histological applications. - Stains nuclei red and cytoplasm pale pink - Contains 80% water, 5% aluminium sulfate, 0.2% nuclear fast red - Enhanced stability - Prevents precipitation over time
The product is subject to the following: Abcam Restricted Use Statement