Learn More
Aat Bioquest Nuclear Green DCS1 5 mM DMSO Solution (0.5 ml)
SDP
Catalog No. 502249741
Encompass_Preferred
Our Nuclear Green DCS1 is a fluorogenic DNA-selective and cell-impermeant dye for analyzing DNA content in dead fixed or apoptotic cells. This catalog is for 0.5 ml.
This item is not returnable. View return policy
Catalog No. 50-224-9741 Supplier Aat Bioquest Supplier No. 17550
Our Nuclear Green DCS1 is a fluorogenic DNA-selective and cell-impermeant dye for analyzing DNA content in dead fixed or apoptotic cells. This catalog is for 0.5 ml.